Masterflex® Series R Valveless Piston Pump Head Adapter Kit
Catalog # MFLX07115-99
Supplier: Avantor Fluid Handling
About this item
Compatible with Masterflex® L/S® or series Q drive.
- No valves to clog or wear out
- Use for low-flow applications
- Kynar® pump body withstands harsh chemicals
- Series R pump heads come equipped with rotating pistons to prevent clogging—ideal for pumping visco
Compression fittings accept 1/4" OD tubing. For a complete assembly, order series R pump head and a Masterflex L/S® or series Q drive. Adapter kits let you attach pump heads to drives.
Flow characteristics may vary with different fluids.
- Description:Series R Pump Head Adapter Kit; Series R Head to L/S Drive
- Seq:01
- Environmentally Preferable:
- Model:07115-99
- Inlet connection:Series R Head to L/S Drive Adapter
- Warranty:1 year
- Pk:1
Frequently Bought Together