Masterflex® L/S® Standard Pump Head, Replacement End Bell Assembly, Polycarbonate Housing, SS Rotor, L/S® 24
Catalog # MFLX07024-91
Supplier: Avantor Fluid Handling
About this item
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- Pump series:L/S
- Description:Standard Pump Head, Replacement End Bell Assembly, Polycarbonate Housing, SS Rotor, L/S® 24 Tubing
- Seq:31
- Environmentally Preferable:
- Model:07024-91
- Open-head sensor:No
- Tubing sizes accepted:L/S 24
- Warranty:2 years
- Pk:1
- Housing material:PC
- Rotor material:Stainless Steel
Frequently Bought Together