ContactUsLinkComponent
Masterflex® Ratchet Tubing Clamp for up to 1/2" Tubing, Acetal; 25/pack
Masterflex® Ratchet Tubing Clamp for up to 1/2" Tubing, Acetal; 25/pack
Artikelnr. MFLX06833-00
undefined
Masterflex® Ratchet Tubing Clamp for up to 1/2" Tubing, Acetal; 25/pack
Artikelnr. MFLX06833-00
About this item
Won't rust, corrode, or distort.
  • Constructed of durable Celcon plastic
Durable Celcon plastic clamp is autoclavable; won't rust, corrode, or distort. Accepts flexible tubing.
Mini clamp accept up to 1/4" OD tubing and provides on/off control. The standard clamp accepts up to 1/2" OD tubing and features twelve-position ratchet control to adjust flow from fully-open to complete-closure.
  • Housing material:
    Polyoxymethylene (POM) Acetal
  • Description:
    Ratchet Tubing Clamp for up to 1/2" Tubing, Acetal; 25/pack
  • Environmentally Preferable:
  • Pinch valve tube size:
    1/8" to 1/2" OD
  • Warranty:
    3 Months
  • Pk:
    25
Frequently Bought Together