ContactUsLinkComponent
Masterflex® Gamma Irradiated Sampling Bottle Assembly, Sterile, PETG Bottle; L
Masterflex® Gamma Irradiated Sampling Bottle Assembly, Sterile, PETG Bottle; L
Catalog # MFLX95644-06
Sterile
CE certificate
undefined
Masterflex® Gamma Irradiated Sampling Bottle Assembly, Sterile, PETG Bottle; L
Catalog # MFLX95644-06
Sterile
CE certificate
  • Special feature:
  • Sterile
  • Sterile
  • Sterile
  • Sterile
  • Sterile
  • Int. Ø tubing:
  • 1/8" ID x 1/4" OD
  • 1/8" ID x 1/4" OD
  • 1/8" ID x 1/4" OD
  • 1/8" ID x 1/4" OD
  • 1/8" ID x 1/4" OD
  • Warranty:
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • Currently Viewing
Specifications
About this item
Safely transfer, sample, and store biopharmaceutical products and reagents.
  • Secure, single-piece cap/tubing assembly eliminates contamination and product loss due to lose connections
  • Assemblies are pre-sterilized by gamma irradiation—eliminates labor and validation costs associated with assembly and cleaning
Cap assembly includes a seamless liner with premolded C-Flex® fill and vent tubes to create an unobstructed fluid path. The 18" fill tube is plugged at the terminal end to maintain sterility until use. Vent tube includes a 0.2-micron PTFE vent filter to prevent infiltration by contaminants. Vent filter is hydrophobic to guard against loss of costly product or media during filling or emptying. The one-piece liner eliminates the risk of bacterial entrapment and contamination and forms an elastomeric seal between cap and bottle rim.
C-Flex tubing is manufactured according to cGMP and meets USP Class VI and USDA requirements. Tubing can withstand temperatures from –100 to 275 °F (–73 to 135 °C). C-Flex tubing has exceptional tensile and tear strength, extremely low extractables, and excellent biocompatibility. The smooth inner surface reduces absorption and protein binding.
Delivery: Includes clear PETG bottle, 53B cap, C-Flex® tube/liner assembly with 1/8" ID x 1/4" OD tubing, and 0.2-micron vent filter.
  • Cap Size:
    53B
  • Capacity:
    1000 ml
  • Description:
    Gamma Irradiated Sampling Bottle Assembly, Sterile, PETG Bottle; L
  • Material:
    Polyethylene terephtalate (PET)
  • Environmentally Preferable:
  • Special feature:
    Sterile
  • Int. Ø tubing:
    1/8" ID x 1/4" OD
  • Warranty:
    3 Months
  • Cap colour:
    Natural
  • Pk:
    1
  • Bottle colour:
    Clear
Frequently Bought Together