PendoTech Single-Use Adapter Tee, 1/2" Hose Barb x 1/2" Hose Barb x Male Luer Fitting, Polycarbonate
Artikelnr. MFLX19406-35
Supplier: Avantor Fluid Handling
CE Compliant
About this item
Optimize performance of your PendoTECH® Pressure Sensor.
- Choose from a wide range of replacement parts and accessories
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
- Max. pressure:76 psi
- Process connection:1/2" x 1/2" hose barb, male luer
- Description:Adapter Tee, 1/2" Hose Barb x 1/2" Hose Barb x Male Luer Fitting, Polycarbonate
- Environmentally Preferable:
- Warranty:3 Months
- Pk:1
Frequently Bought Together