Orders LinkContactUsLinkComponent
Masterflex® Sanitary Gaskets, Polymers, Avantor®
Masterflex® Sanitary Gaskets, Polymers, Avantor®
Sanitary Gasket, Viton®, 3/4" Tri-Clamp®; 10/PK, Viton®, 3/4", 3/4"
Catalog # MFLX30548-20
CE certificate
Masterflex® Sanitary Gaskets, Polymers, Avantor®
Catalog # MFLX30548-20
CE certificate
Masterflex® Sanitary Gaskets, Polymers, Avantor®
Masterflex® Sanitary Gaskets, Polymers, Avantor®
Sanitary Gasket, Viton®, 3/4" Tri-Clamp®; 10/PK, Viton®, 3/4", 3/4"
Price
51,70 €
Log in for availability
1 * 10 ST
Product Details & Documents
Ensure a tight seal.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimise downtime
These gaskets are for Tri-Clamp® style fittings and come in a wide variety of materials to match specific process needs. All gaskets meet USDA, 3-A and CGMP criteria. FDA Compliant. All silicone, EPDM, PTFE, and Viton® gaskets are animal-derived ingredient free and meet USP Class VI and cytotoxicity criteria.
Platinum-cured silicone is ideal for most pharmaceutical, food, and beverage processing applications. Odorless and tasteless to ensure media purity. May undergo SIP. Usable from –58 to 446 °F (–50 to 230 °C).
Viton offers excellent chemical resistance, especially to many acids and alkalis.
Specifications
  • Sanitary clamp size (Port 1):
    3/4"
  • Description:
    Sanitary Gasket, Viton®, 3/4" Tri-Clamp®; 10/PK
  • Seq:
    31
  • Fitting material:
    Viton®
  • Sanitary gasket size:
    3/4"
  • Cat. No.:
    MFLX30548-20
  • Warranty:
    1 year
  • Connection type:
    Sanitary