Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Express Eingabe
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MB4 BUFFER 3X1000ML 1 * 3 L
Catalog # OMEGMB4-3000
Supplier:
OMEGA BIO-TEK
Bild vergrößern
MB4 BUFFER 3X1000ML 1 * 3 L
Catalog # OMEGMB4-3000
Supplier:
OMEGA BIO-TEK
Frequently Bought Together
Restriction enzymes, BamHI, HC, Fermentas
Supplier: Thermo Fisher Scientific
ASSAYCOMPLETE CELL PLATING 21 REAGEN 1 * 500 mL
Supplier: DISCOVERX