Order Entry
Austria
Orders LinkContactUsLinkComponent
Urea

Urea

Poly pail, 12 kg
Catalog # SPCMUR107-12KG
Supplier:   Spectrum Chemical
Urea
Catalog # SPCMUR107-12KG
Supplier:  Spectrum Chemical
CAS Number:  57-13-6
Urea

Poly pail, 12 kg

CAS Number:  57-13-6
Price
1 640,00 €
Log in for availability
1 * 12 kg
Product Details & Documents

Urea, USP, JP is used in topical dermatological products to promote rehydration of the skin. Urea preparations are also used for non-surgical removal of damaged or dead nails. All Spectrum Chemical USP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Synonyms:  Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
  • CAS Number:57-13-6
  • Formula:NH₂CONH₂
  • MDL Number:MFCD00008022
  • Molecular Weight:60.06 g/mol

Documents

Specifications

Specifications

  • Pack type:
    Poly pail
  • Pk:
    12 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.