WIPER MIRAWIPE 11INX11IN 1 * 25 items
Catalog # FOAMHT5711-25
Supplier: FOAMTEC
Frequently Bought Together
Accessories for dissolved oxygen probes, IDS FDO®
Supplier: WTW
Sampling swabs, Alpha®
Supplier: Texwipe
Paper towel rolls, zetRoll®
Supplier: Zellstoff-Vertriebs-GmbH & Co. KG
Wipes for general applications, WYPALL* L40
Supplier: KIMBERLY CLARK
Wipe rolls, Multiclean®
Supplier: Zellstoff-Vertriebs-GmbH & Co. KG
Cleanroom wipes, pre-saturated, meltblown polypropylene, PROSAT®
Supplier: CONTEC
Wipe rolls, zetRoll®
Supplier: Zellstoff-Vertriebs-GmbH & Co. KG
VWR®, Dispenser for Wipe Rolls
Supplier: VWR Collection
Reference electrode, half cell, Orion™ 900200 Sure-Flow™
Supplier: Thermo Orion
Economy pH electrodes, InLab® Easy
Supplier: Mettler - Toledo
TISSUE TOILETPAPER WHITE 3LAY-250SH/ROLL 1 * 72 Roll
Supplier: Zellstoff-Vertriebs-GmbH & Co. KG
CLEANING CLOTH MULTITEX Z47 WHITE 1 * 1 Roll
Supplier: Zellstoff-Vertriebs-GmbH & Co. KG
pH combination electrodes, IDS®
Supplier: SI Analytics
Microfibre Cloths
Supplier: Rubbermaid Commercial Products
VWR® B'Wipes 5000, Cleanroom wipes
Supplier: VWR Collection
pH electrodes for fast results, InLab®
Supplier: Mettler - Toledo
TORK XPRESS SOFT MULTIF. TOWEL 21X150PCS 1 * 3.150 items
Supplier: Essity
ORP combination electrodes, IntelliCAL™ MTC101 / MTC301
Supplier: Hach