METHYLENE BLUE RAL 555 AQUEOUS SOL. 2.5L 1 * 2,5 L
Artikelnr. 720-0358
Supplier: RAL
Frequently Bought Together
Hydrochloric acid 32%, EMSURE® for analysis, Supelco®
Supplier: Merck
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Water, Molecular Biology Grade
Supplier: PanReac AppliChem
FIX RAL 555 2.5L 1 * 2,5 L
Supplier: RAL
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Polyethylene glycol 400, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Methyl blue microbiology stain (C.I. 42780)
Supplier: VWR Chemicals