1GR PSDVB NIST 1000UM 1000 UM, PACKAGED IN 15 ML 1 * 1 g
Catalog # MSPP4400A
Supplier: MARKET SOURCE PART PROCESS
1GR PSDVB NIST 1000UM 1000 UM, PACKAGED IN 15 ML 1 * 1 g
Catalog # MSPP4400A
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Transfer pipettes
Supplier: RATIOLAB
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Serological pipettes
Supplier: Greiner Bio-One
Chloroform, LiChrosolv® for HPLC
Supplier: Merck
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Cryoboxes, 136×136 mm
Supplier: RATIOLAB
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
Supplier: VWR Collection
VWR®, Aluminium Foil Seals for PCR and Storage (96- and 384-Well Plates)
Supplier: VWR Collection
Biotube™ system
Supplier: Simport Scientific
Methanol ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Cellulose chromatography papers, Whatman™
Supplier: Whatman products (Cytiva)
Collagen I culture dishes, Corning® BioCoat™
Supplier: Corning
Cryoboxes, coloured, Nalgene®
Supplier: Thermo Fisher Scientific
Accessories and consumables for KingFisher™ Flex
Supplier: Thermo Fisher Scientific