Tubing, PVC, clear 7x10mm thickness 1.5mm roll lenth 100m. 1 * 100 m
Catalog # DENE3500710B
Supplier: DEUTSCH NEUMANN
Tubing, PVC, clear 7x10mm thickness 1.5mm roll lenth 100m. 1 * 100 m
Catalog # DENE3500710B
Supplier: DEUTSCH NEUMANN
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® ION-15A, Ioniser for Neutralising Electrostatic Build-Up in Weighing Samples
Supplier: VWR Collection
Tubing connectors, straight
Supplier: KARTELL
VWR®, Safety Pipette Fillers, Standard/Universal/Flip
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Sulfonation flasks, with three necks
Supplier: witeg Labortechnik
Metric non-phthalate PVC tubing, Nalgene™
Supplier: Thermo Fisher Scientific
Liebig condenser with standard ground joints
Supplier: SciLabware
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Straight, HDPE, Avantor®
Supplier: Avantor Fluid Handling