ContactUsLinkComponent
FILTER ASSY, EMISSION 580-630nm: EtBr, ELITE MOUNT 1 * 1 items
Catalog # UVPA38-0220-05
FILTER ASSY, EMISSION 580-630nm: EtBr, ELITE MOUNT 1 * 1 items
Catalog # UVPA38-0220-05
Frequently Bought Together