[EN]REFRACTOMETER HAND 0-40 BRIX 0-25% V 1 * 1 ST
Catalog # EUMXRF.6627
Supplier: EUROMEX
Frequently Bought Together
Camera adapters
Supplier: MOTIC
Accessories for flow cups with interchangeable nozzle
Supplier: Industrial Physics
VWR®, Combs for PerfectBlue™ Gel System Maxi M
Supplier: VWR Collection
REFRACTOMETER GLYCOL 1 * 1 items
Supplier: Lovibond
Accessories for chromatography systems
Supplier: Cytiva
Components for spectrophotometric colorimeters, PFX<i>i</i>-195/880/950/995 series, Lovibond®
Supplier: Lovibond Tintometer
VWR®, Eluent buffers for amino acid analyzer
Supplier: VWR Chemicals
VWR®, Bead Mill Homogeniser
Supplier: VWR Collection
[EN]BIOSENSOR/ANTI-MURINE IGG FV TRAY
Supplier: PALL FORTEBIO
Storage phosphor screens and cassettes
Supplier: Cytiva
CAMERA HB 1080P 2MP CMOS HDMI 1 * 1 items
Supplier: OPTIKA
Accessories for chloride analyser, model 926
Supplier: SHERWOOD SCIENTIFIC INSTRUMENT