ContactUsLinkComponent
The Katana Lab probe works on Attenuated Total Reflection (ATR) principle Possible Applications: Determination of color numbers,Analysis of colorants and dyes) 1 * 1 items
Catalog # HELLPK1KSSZUEASM24
Supplier:  HELLMA
The Katana Lab probe works on Attenuated Total Reflection (ATR) principle Possible Applications: Determination of color numbers,Analysis of colorants and dyes) 1 * 1 items
Catalog # HELLPK1KSSZUEASM24
Supplier:  HELLMA
Frequently Bought Together