TSX Series High-Performance Lab Refrigerator 827 L 3-7C (5C) 1 * 1 items
Catalog # REVCTSX3005SV
Supplier: REVCO TECHNOLOGIES
TSX Series High-Performance Lab Refrigerator 827 L 3-7C (5C) 1 * 1 items
Catalog # REVCTSX3005SV
Supplier: REVCO TECHNOLOGIES
Frequently Bought Together
VWR® Traceable® Calibrated Timer
Supplier: VWR Collection
Repetitive pipette, Multipette® M4
Supplier: EPPENDORF
Single-channel pipettes, mechanical, fixed / variable volume, Finnpipette™ F2
Supplier: Thermo Fisher Scientific
3-Packs, Research® plus (General Lab Product)
Supplier: EPPENDORF
VWR® MiniStar/MiniStar blueline, Microcentrifuge
Supplier: VWR Collection
Pipette controller, PIPETBOY acu 2
Supplier: Integra
VWR® Pipette Stands
Supplier: VWR Collection
VWR® Traceable® Digital Thermometer
Supplier: VWR Collection
VWR®, Digital Timer, Four Channels, with White Board
Supplier: VWR Collection
Wall clock, classic
Supplier: MOINEAU
Eppendorf® Pipette Holder System
Supplier: EPPENDORF
VWR®, Digital Timer, Four Channels, with Clock
Supplier: VWR Collection
VWR®, Digital Timer, One Channel
Supplier: VWR Collection
Accessories for marco
Supplier: Brand
Cartridges for Milli-Q®
Supplier: Merck
TRAY - SINGLE WAFER 100MMH22-40-0615 1 * 100 ST
Supplier: ENTEGRIS