Order Entry
Austria
Orders LinkContactUsLinkComponent
Nitrile gloves, Kimtech™ Opal™

Nitrile gloves, Kimtech™ Opal™

M, Dark blue, 240 mm, 200/box
Catalog # 112-6225
   Waste Reducing
Supplier:   Ansell
Nitrile gloves, Kimtech™ Opal™
Catalog # 112-6225
Supplier:  Ansell
   Waste Reducing
Nitrile gloves, Kimtech™ Opal™
Nitrile gloves, Kimtech™ Opal™

M, Dark blue, 240 mm, 200/box

Price
37,70 €
Log in for availability
1 * 200 ST
Product Details & Documents

Kimtech™ Opal™ nitrile gloves guard against contamination by chemical splash and microorganism hazards. Powder-free, ambidextrous, with beaded cuff and textured fingertips. The absence of natural rubber latex and vulcanising agents reduces the risk of TYPE I and IV glove associated skin reactions.

  • Assured compliance
  • Formulated without thiurams, thiazoles, guanidines, carbamates, and no sulphur added
  • Soft, comfortable, and easy to don
  • Textured fingertips enhance grip and tactile sensitivity for safer and more efficient processes
  • Dark colour to enhance concentration on task
  • Available in a wide range of sizes, from XS to XL

These gloves guard against contamination by chemical splash and microorganism hazards, delivering seamless protection when and where it counts. Powder-free gloves that are ideal for use in research and production facilities, life sciences, and non sterile drug manufacturing applications. The gloves feature an innovative approach using a thin nitrile polymer with a tip thickness of just 0,07 mm, but with excellent tensile strength suitable for rigorous process use. Easy to don, soft and comfortable, they are suitable for extended wearing time.

The absence of natural rubber latex, sulphur and vulcanising agents reduces the risk of TYPE I and TYPE IV glove associated skin reactions, protecting the wearer as well as the application. Precision manufacturing reduces the risk of contamination and ensures the nitrile gloves offer high levels of regulatory compliance. Beaded cuffs and textured fingertips enable excellent handling of both wet and dry materials, and the ambidextrous gloves have been extensively tested under stringent conditions. In addition, the thin nitrile construction and efficient packaging mean that the chemical resistant gloves can reduce waste by up to 50%.

Certifications: EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS), Cat. III; AQL 1,5
Food contact approved

Documents

Specifications
  • Glove size:
    M
  • Packed:
    200/box
  • Colour:
    Dark blue
  • Length:
    240 mm
  • Seq:
    03
  • Cat. No.:
    112-6225
  • Latex / latex-free:
    Latex-free
  • Food contact approved:
    +
  • Disposable / reusable:
    Disposable
  • Material:
    Nitrile
  • Cuff style:
    Beaded
  • Powdered / powder-free:
    Powder-free
  • Environmentally Preferable:
    Waste Reducing
  • EN norm:
    EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS)
  • Finish:
    Textured fingertips
  • Ambidextrous / hand-specific:
    Ambidextrous