ContactUsLinkComponent
hose MT-140-1,5-G3/4 stainless steel, 1.4404 with Braiding Temperature range: -40°C…140°C Length: 150 cm Connection: G3/4 Max. pressure at 20°C: 10 bar Internal diameter (DN): 19 1 * 1 items
Catalog # HUBE10810
Supplier:  HUBER
hose MT-140-1,5-G3/4 stainless steel, 1.4404 with Braiding Temperature range: -40°C…140°C Length: 150 cm Connection: G3/4 Max. pressure at 20°C: 10 bar Internal diameter (DN): 19 1 * 1 items
Catalog # HUBE10810
Supplier:  HUBER
Frequently Bought Together