ContactUsLinkComponent
Epstein-Barr virus (Herpesvirus 4) EBV Glycoprotein gp350 Protein, His Tag (GP0-E52H6) is expressed from human 293 cells (HEK293). I t contains AA Met 1 - Thr 470 (Accession # P03200-1). 1 * 1 mg
Catalog # ACRBGP0-E52H6-1MG
Supplier:  ACROBIOSYSTEMS
Epstein-Barr virus (Herpesvirus 4) EBV Glycoprotein gp350 Protein, His Tag (GP0-E52H6) is expressed from human 293 cells (HEK293). I t contains AA Met 1 - Thr 470 (Accession # P03200-1). 1 * 1 mg
Catalog # ACRBGP0-E52H6-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together