ContactUsLinkComponent
Human Angiopoietin-2, His Tag (AN2-H52H4) is expressed from human 293 cells (HEK293). It contains AA Tyr 19 - Phe 496 (Accession # O 15123-1). 1 * 1 mg
Catalog # ACRBAN2-H52H4-1MG
Supplier:  ACRO Biosystems
Human Angiopoietin-2, His Tag (AN2-H52H4) is expressed from human 293 cells (HEK293). It contains AA Tyr 19 - Phe 496 (Accession # O 15123-1). 1 * 1 mg
Catalog # ACRBAN2-H52H4-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together