ContactUsLinkComponent
Biotinylated Cynomolgus TSLP, His,Avitag (TSP-C82E3) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Ac cession # A0A7N9CAT7-1). 1 * 200 µG
Catalog # ACRBTSP-C82E3200UG
Supplier:  ACRO Biosystems
Biotinylated Cynomolgus TSLP, His,Avitag (TSP-C82E3) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Ac cession # A0A7N9CAT7-1). 1 * 200 µG
Catalog # ACRBTSP-C82E3200UG
Supplier:  ACRO Biosystems
Frequently Bought Together