ContactUsLinkComponent
Human IFN-alpha 2b (K46R), Fc Tag (IFB-H5253) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Glu 188 (Accession # P01563-1 (K46R)). 1 * 100 µG
Catalog # ACRBIFB-H5253100UG
Supplier:  ACRO Biosystems
Human IFN-alpha 2b (K46R), Fc Tag (IFB-H5253) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Glu 188 (Accession # P01563-1 (K46R)). 1 * 100 µG
Catalog # ACRBIFB-H5253100UG
Supplier:  ACRO Biosystems
Frequently Bought Together