ContactUsLinkComponent
Mouse IL-2 R gamma, His Tag (ILA-M52H3) is expressed from human 293 cells (HEK293). It contains AA Trp 23 - Ala 263 (Accession # P34 902-1). 1 * 100 µG
Catalog # ACRBILA-M52H3100UG
Supplier:  ACRO Biosystems
Mouse IL-2 R gamma, His Tag (ILA-M52H3) is expressed from human 293 cells (HEK293). It contains AA Trp 23 - Ala 263 (Accession # P34 902-1). 1 * 100 µG
Catalog # ACRBILA-M52H3100UG
Supplier:  ACRO Biosystems
Frequently Bought Together