Human IL-17A&IL-17F Heterodimer Protein, Twin Strep&His Tag (ILF-H52W6) is expressed from human 293 cells (HEK293). It contains AA G ly 24 - Ala 155 (IL-17A) & Arg 31 - Gln 163 (IL-17F) (Accession # Q16552-1 (IL-17A) & Q96PD4-1 (IL-17F)). Two sequent 1 * 100 µG
Catalog # ACRBILF-H52W6100UG
Supplier: ACRO Biosystems
Frequently Bought Together