ContactUsLinkComponent
Mouse NKG2A, His Tag (NKA-M52H5) is expressed from human 293 cells (HEK293). It contains AA Thr 95 - Ile 244 (Accession # Q9Z202-1). 1 * 1 mg
Catalog # ACRBNKA-M52H5-1MG
Supplier:  ACRO Biosystems
Mouse NKG2A, His Tag (NKA-M52H5) is expressed from human 293 cells (HEK293). It contains AA Thr 95 - Ile 244 (Accession # Q9Z202-1). 1 * 1 mg
Catalog # ACRBNKA-M52H5-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together