ContactUsLinkComponent
Mouse IL-5, His Tag (IL5-M52H3) is expressed from human 293 cells (HEK293). It contains AA Met 21 - Gly 133 (Accession # P04401-1). 1 * 50 µG
Catalog # ACRBIL5-M52H3-50UG
Supplier:  ACRO Biosystems
Mouse IL-5, His Tag (IL5-M52H3) is expressed from human 293 cells (HEK293). It contains AA Met 21 - Gly 133 (Accession # P04401-1). 1 * 50 µG
Catalog # ACRBIL5-M52H3-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together