ContactUsLinkComponent
Human USAG-1, Fc Tag (US1-H5254) is expressed from human 293 cells (HEK293). It contains AA Phe 24 - Ser 206 (Accession # Q6X4U4-1). 1 * 100 µG
Catalog # ACRBUS1-H5254100UG
Supplier:  ACRO Biosystems
Human USAG-1, Fc Tag (US1-H5254) is expressed from human 293 cells (HEK293). It contains AA Phe 24 - Ser 206 (Accession # Q6X4U4-1). 1 * 100 µG
Catalog # ACRBUS1-H5254100UG
Supplier:  ACRO Biosystems
Frequently Bought Together