CytoFLEX Startup Reagents and Accessories for systems without the IR Laser. Includes Daily QC Fluorospheres, Sheath fluid, FlowClean, Contrad 70 concentrate and sample tubes. 1 * 1 KIT
Catalog # BECMB55031
Supplier: Beckman Coulter
Frequently Bought Together