ContactUsLinkComponent
Each impact-resistant 50 mL tube contains 0.5 mm diameter Yttria-Stabilized Zirconium Oxide beads. Y2O3 + ZrO2 has high density and has extremely high hardness making this matrix ideal for cracking of very tough cell walls, while it's smooth 1 * 10 items
Catalog # 116979010.
Supplier:  MP Biomedicals
Each impact-resistant 50 mL tube contains 0.5 mm diameter Yttria-Stabilized Zirconium Oxide beads. Y2O3 + ZrO2 has high density and has extremely high hardness making this matrix ideal for cracking of very tough cell walls, while it's smooth 1 * 10 items
Catalog # 116979010.
Supplier:  MP Biomedicals
Frequently Bought Together