Measured by its binding ability in a functional ELISA. Immobilized Recombinant SARS-CoV-2 Spike S1 at 2 ug/mL (100 uL/well) can bind recombinant Human ACE2 with a linear range of 0.15-6.85 ng/mL. Immobilized Human ACE2 on COOH Chip, can bind SARS-COV 1 * 100 µG
Catalog # AVIVOPAN06665
Supplier: Aviva Systems Biology
Frequently Bought Together