Orders LinkContactUsLinkComponent
Watch glasses
Watch glasses
Watch glass, glass, with fused rim, 120 mm
Catalog # HECH40010012
Watch glasses
Catalog # HECH40010012
Watch glasses
Watch glasses
Watch glass, glass, with fused rim, 120 mm
Price
19,70 €
Log in for availability
1 * 10 ST
Product Details & Documents
Evaporating dishes, made of glass, with fused rim.
  • Good chemical restistance
  • Available with or without hole
Certifications: DIN 12 341
Specifications
  • Ø:
    120 mm
  • Description:
    Watch glass, glass, with fused rim
  • Seq:
    18