ContactUsLinkComponent
Micro-Sample Management System.This complete Set includes 96, 350ul RSA-Pro X Glass Inserts in a Rack and Base and a Solvent Resist ant Polymeric Plug Style Sealing Mat. U-2D Brand. 1 EA. 1 * 1 items
Catalog # MICS95296-350-1PR
Supplier:  MICROSOLV
Micro-Sample Management System.This complete Set includes 96, 350ul RSA-Pro X Glass Inserts in a Rack and Base and a Solvent Resist ant Polymeric Plug Style Sealing Mat. U-2D Brand. 1 EA. 1 * 1 items
Catalog # MICS95296-350-1PR
Supplier:  MICROSOLV
Frequently Bought Together