TDB-120, Dry-block Thermostat with block,,built-in A-103 block** for 21x 0.5 ml + 32 x 1.5 ml + 50 x 0.2 ml microtubes 1 * 1 items
Catalog # BSANBS-010401-QAA
Supplier: BIOSAN
TDB-120, Dry-block Thermostat with block,,built-in A-103 block** for 21x 0.5 ml + 32 x 1.5 ml + 50 x 0.2 ml microtubes 1 * 1 items
Catalog # BSANBS-010401-QAA
Supplier: BIOSAN
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection