Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Medical mask Op-Air, earloop EN 14683 Type II - Made in France - white 1 * 960 items
Artikelnr. HOPEM14301-30
Supplier:
Medicom
Expand image
Medical mask Op-Air, earloop EN 14683 Type II - Made in France - white 1 * 960 items
Artikelnr. HOPEM14301-30
Supplier:
Medicom
Related Information
Dokumentation
Datasheet
TDS Medical Face Mask with Earloops.pdf
Dokumentation
Frequently Bought Together
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
Medical Masks, Type IIR
Supplier: Medicom
VWR® Nitrile Extra Light, Nitrile Gloves
Supplier: VWR Collection
VWR®, Laboratory Markers
Supplier: VWR Collection
Nitrile gloves, TouchNTuff® 92-600 / 92-605
Supplier: Ansell
Gloves, nitrile/neoprene, Microflex® 93-260
Supplier: Ansell
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR® Labels, Hazardous Substance, GHS Labels
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Latex Gloves
Supplier: VWR Collection