ContactUsLinkComponent
Safety cabinet with 1 wing door (right hand) for the storage of acids and lyes in clean rooms. Outer dimensions: 595x595x1980 (WxDxH) mm, Inner dimensions: 536x507x1895 (WxDxH) mm, Outer corpus in powder-coated sheet steel, silver. Inner corpus in po 1 * 1 items
Catalog # DUEH20-200667-006
Supplier:  DUPERTHAL
Safety cabinet with 1 wing door (right hand) for the storage of acids and lyes in clean rooms. Outer dimensions: 595x595x1980 (WxDxH) mm, Inner dimensions: 536x507x1895 (WxDxH) mm, Outer corpus in powder-coated sheet steel, silver. Inner corpus in po 1 * 1 items
Catalog # DUEH20-200667-006
Supplier:  DUPERTHAL
Frequently Bought Together