ContactUsLinkComponent
analogue weather station out of stainless steel with barometer, thermometer and hygrometer weather proof, with attachment for hanging, dimensions 96 x 35 x 282mm, Made in Germany 1 * 1 items
  HECH43662010
analogue weather station out of stainless steel with barometer, thermometer and hygrometer weather proof, with attachment for hanging, dimensions 96 x 35 x 282mm, Made in Germany 1 * 1 items
  HECH43662010
Frequently Bought Together