ContactUsLinkComponent
MATTEK 35MM DISH, UNCOATED, COVERSLIP NO. 1.5, 10MM GLASS DIAMETER 1 * 200 ST
Catalog # MSPPP35G-1.5-10-C
MATTEK 35MM DISH, UNCOATED, COVERSLIP NO. 1.5, 10MM GLASS DIAMETER 1 * 200 ST
Catalog # MSPPP35G-1.5-10-C
Frequently Bought Together