Dimethenamid-Oxalamid 1 * 25 mg
Catalog # EHERC12677540
Supplier: EHRENSTORFER
Frequently Bought Together
VWR®, Osmolality standards
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Conductivity standards, Certipur® Supelco®
Supplier: Merck
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR® PESTINORM®, Tetrahydrofuran (THF), Secondary Reference Standard for GC
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, pH-Indicator Paper Congo Red Reag. Ph. Eur. 1022002
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
Certified Reference Materials for environmental analysis
Supplier: VKI WATER QUALITY INSTITUTE
VWR®, Sucrose (Brix) standards, stabilised solutions for refractometry
Supplier: VWR Chemicals
Buffer solutions
Supplier: REAGECON
VWR®, Chloride 1,000 mg/L in Water standard for ion chromatography
Supplier: VWR Chemicals
pH buffers, for LAQUAtwin meters
Supplier: HORIBA
pH buffer, single-use, SINGLET™
Supplier: Hach
N,N-Dimethylsulfamide certified 1 * 100 mg
Supplier: EHRENSTORFER