Human IL-10 ELISPOT Set
Human IL-10 ELISPOT Set
Catalog # ABCAAB46549-10X96
Supplier: Abcam
Jump to:
Product Details & Documents
Human IL-10 ELISPOT Set is a ELISPOT Set for the sensitive detection of Human IL-10 production in a single cell suspension.
Specifications
Specifications
- Description:Human IL-10 ELISPOT Set
- Cat. No.:ABCAAB46549-10X96
- Sample type:Suspension cells
- Form:Uncoated
- Species reactivity:Human
- Storage temperature:4 °C
- Processing time:Multiple steps
- Regulatory status:RUO
- Antigen symbol:IL-10
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



