Human IL-10 ELISPOT Set
Human IL-10 ELISPOT Set
Catalog # ABCAAB46549-15X96
Supplier: Abcam
Jump to:
Product Details & Documents
Human IL-10 ELISPOT Set is a ELISPOT Set for the sensitive detection of Human IL-10 production in a single cell suspension.
Specifications
Specifications
- Description:Human IL-10 ELISPOT Set
- Cat. No.:ABCAAB46549-15X96
- Sample type:Suspension cells
- Form:Uncoated
- Species reactivity:Human
- Storage temperature:4 °C
- Processing time:Multiple steps
- Regulatory status:RUO
- Antigen symbol:IL-10



