ContactUsLinkComponent
The testo 915i wireless thermometer with fast-response air probe and sprung thermocouple strip will enable you to tackle your temperature measurements with greater speed, flexibility and ease, even when dealing with uneven surfaces. The testo 915i au 1 * 1 items
Catalog # TEST0563.2915
Supplier:  Testo
The testo 915i wireless thermometer with fast-response air probe and sprung thermocouple strip will enable you to tackle your temperature measurements with greater speed, flexibility and ease, even when dealing with uneven surfaces. The testo 915i au 1 * 1 items
Catalog # TEST0563.2915
Supplier:  Testo
Frequently Bought Together