REFRACTOMETER STANDARD 50% BRIX 1 * 10 mL
Catalog # HANNHI4020-11
Supplier: Hanna
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
pH buffer solutions, certified (DKD)
Supplier: Mettler - Toledo
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Buffer solutions
Supplier: REAGECON
Griseofulvin certified 1 * 0,25 g
Supplier: EHRENSTORFER
VWR®, Density standards referenced at 15 °C
Supplier: VWR Chemicals
MYCOPLASMA PNEUMONIAE PCR QUANTISTANDARD 1 * 1 ST
Supplier: MARKET SOURCE PART PROCESS
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond
Sulfite lr tablets in pack of 100 1 * 100 Tablet
Supplier: Lovibond
Supelco® Organic Reference Standard, Naphthalene-d₈ solution
Supplier: Merck
Alcohol calibration standards, Ethanol, (CRM), Supelco®
Supplier: Merck
Turbidity calibration standards, formazin, Supelco®
Supplier: Merck