ContactUsLinkComponent
20 Micron Porous Filters, UHMW Polyethylene, 1/8IN (3.2mm) ID, Erweka compatible 1 * 100 items
Catalog # QLAAFIL020-EW-A
20 Micron Porous Filters, UHMW Polyethylene, 1/8IN (3.2mm) ID, Erweka compatible 1 * 100 items
Catalog # QLAAFIL020-EW-A
Frequently Bought Together