ContactUsLinkComponent
WNT7A ELISA Kit (Human)is based on standard sandwich enzyme-linked immuno-sorbent assay technology. An antibody specific for WNT7A has been pre-coated onto a 96-well plate (12 x 8 Well Strips) and blocked. Standards or test samples are added to the w 1 * 1 KIT
Catalog # AVIVOKEH01755
WNT7A ELISA Kit (Human)is based on standard sandwich enzyme-linked immuno-sorbent assay technology. An antibody specific for WNT7A has been pre-coated onto a 96-well plate (12 x 8 Well Strips) and blocked. Standards or test samples are added to the w 1 * 1 KIT
Catalog # AVIVOKEH01755
Frequently Bought Together