Orders LinkContactUsLinkComponent
Label maker, b.safe
Label maker, b.safe
Portable label printer
Catalog # 590-0217
Supplier:   Bohlender
Label maker, b.safe
Catalog # 590-0217
Supplier:  Bohlender
Label maker, b.safe
Label maker, b.safe
Portable label printer
Price
123,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Portable label printer with computer-style keyboard, large LCD display and 20 methods to format text.
  • Portable
  • Powered batteries or optional AC-adapter
Powered by 6 batteries AAA or optional AC-adapter (both not included).
Delivery information: Includes one label cassette black on white, width 12 mm/length 3 m. Labels up to a width of 12 mm can be printed.
Specifications
  • Description:
    Portable label printer
  • Seq:
    01