Tris(hydroxymethyl)aminomethane hydrochloride EMPROVE EXPERT 1 * 5 kg
Catalog # 1.08219.5000
Supplier: MERCK PRODUCTION CHEMICALS
Tris(hydroxymethyl)aminomethane hydrochloride EMPROVE EXPERT 1 * 5 kg
Catalog # 1.08219.5000
Supplier: MERCK PRODUCTION CHEMICALS
Frequently Bought Together
2,2,4-Trimethylpentane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
tert-Butyl methyl ether, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
Tetrahydrofuran, EMPLURA®, extra pure, Supelco®
Supplier: Merck
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
n-Hexane ≥99%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Masterflex® Union Fittings, Hose Barb, Straight, Polypropylene, Avantor®
Supplier: Avantor Fluid Handling