ContactUsLinkComponent
The biotinylated CD33 protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can capture the anti-CD33 antibody from various assay systems. The beads are in uniform size, narrow size distribution with large surf 1 * 10 mg
Catalog # ACRBMBC-K008-10MG
Supplier:  ACRO Biosystems
The biotinylated CD33 protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can capture the anti-CD33 antibody from various assay systems. The beads are in uniform size, narrow size distribution with large surf 1 * 10 mg
Catalog # ACRBMBC-K008-10MG
Supplier:  ACRO Biosystems
Frequently Bought Together