ContactUsLinkComponent
Paguma larvata ACE2, His Tag AC2-P5248 is expressed from human 293 cells HEK293. It contains AA Gln 18 - Thr 740 Accession Q56NL1-1 . 1 * 1 mg
Catalog # ACRBAC2-P5248-1MG
Supplier:  ACRO Biosystems
Paguma larvata ACE2, His Tag AC2-P5248 is expressed from human 293 cells HEK293. It contains AA Gln 18 - Thr 740 Accession Q56NL1-1 . 1 * 1 mg
Catalog # ACRBAC2-P5248-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together