ContactUsLinkComponent
FITC-Labeled Human CD72, Fc Tag CD2-HF258 is expressed from human 293 cells HEK293. It contains AA Arg 117 - Asp 359 Accession P218 54-1. It is the FITC labeled form of Human CD72, Fc Tag Cat. No.CD2-H5251. 1 * 25 µG
Catalog # ACRBCD2-HF258-25UG
Supplier:  ACROBIOSYSTEMS
FITC-Labeled Human CD72, Fc Tag CD2-HF258 is expressed from human 293 cells HEK293. It contains AA Arg 117 - Asp 359 Accession P218 54-1. It is the FITC labeled form of Human CD72, Fc Tag Cat. No.CD2-H5251. 1 * 25 µG
Catalog # ACRBCD2-HF258-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together