ContactUsLinkComponent
Cynomolgus CD32b, His Tag CDB-C5224 is expressed from human 293 cells HEK293. It contains AA Ala 46 - Pro 224 Accession Q8SPW3-1. 1 * 100 µG
Catalog # ACRBCDB-C5224100UG
Supplier:  ACRO Biosystems
Cynomolgus CD32b, His Tag CDB-C5224 is expressed from human 293 cells HEK293. It contains AA Ala 46 - Pro 224 Accession Q8SPW3-1. 1 * 100 µG
Catalog # ACRBCDB-C5224100UG
Supplier:  ACRO Biosystems
Frequently Bought Together